Learn More
StatLab™ Prostate Biopsy Fixative with Eosin
GSA_VASDP
Catalog No. PF0507B500
Change view
Click to view available options
Quantity:
500 x 20 mL
Catalog No. Quantity
PF0507B500 500 x 20 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. PF0507B500 Supplier StatLab™ Supplier No. PF0507B500
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
StatClick™ Vials filled with a special formulation for prostate biopsies.
  • Has slight Eosin tint to ensure easy identification of biopsies after processing and embedding
  • StatClick™ patented vials produce an audible 'clicking' sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Packaging Type Vial
Quantity 500 x 20 mL
Formulation Formalin and Alcohol