Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Prostate Biopsy Kit 10% NBF
GSA_VASDP

Catalog No. STLPBKNB12
Change view
Click to view available options
No. per Pack:
12 Pk.
16 Pk.
6 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
STLPBKNB12 12 Pk.
STLPBKNB6 6 Pk.
STLPBKNB16 16 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLPBKNB12 Supplier StatLab™ Supplier No. PBKNB12
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

Pre-labeled kit with location diagram contains prefilled NBF arranged in pre-labeled positions that correlate with the area of the prostate from which sample is taken.

  • Biopsy location stickers included
  • StatClick Vials are filled with 10% Neutral Buffered Formalin

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Biopsy Container
Closure Included Yes
Graduated Yes
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 20 mL
No. per Pack 12 Pk.
Fit for Purpose Certified Quality
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.