Learn More
StatLab Prostate Biopsy Kit
Description
- Kit contains prefilled StatClick vials with 10% neutral buffered formalin solution arranged in pre-labeled positions that correlate with the area of the prostate from which sample is taken, and stickers to affix to the prefill indicating the biopsy location
- 8-vial biopsy kit with StatClick patented vials produce an audible clicking sound when the leak-proof seal has been properly engaged assuring accurate closure and maintaining specimen integrity during transport
- 1 kit
Specifications
Specifications
| Product Type | Pre-filled Formalin Container |
| Color | Clear |