Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostate Biopsy Kits with Neutral Buffered Formalin, StatLab
SDP

Catalog No. 22035377
Change view
Click to view available options
Quantity:
14 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-035-377 14 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-035-377 Supplier SLMP Llc Dba Statlab Medical Supplier No. PBKNB14
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Prostate Biopsy Kits with Neutral Buffered Formalin - 14 vial kit.

  • Kit contains prefilled StatClick vials with 10% neutral buffered formalin solution arranged in pre-labeled positions that correlate with the area of the prostate from which sample is taken, and stickers to affix to the prefill indicating the biopsy location
  • Pre-labeled with a location diagram supporting accurate collection

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 14 Vials
Solution Type Formalin
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.