Learn More
Discovery Life Sciences Prostate Cancer Active Tx PBMC
SDP
Catalog No. 5000803995
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-03995 Supplier Discovery Life Sciences Supplier No. 120160
Add to Cart
Add to Cart
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays