Learn More
ATCC Prostate Epithelial Cell Growth Kit
Catalog No. 502383922
Encompass
Prostate Epithelial Cell Growth Kit (ATCC PCS-440-040) contains components that when added to Prostate Epithelial Cell Basal Medium (ATCC PCS-440-030) create a complete ATCC Primary Cell Solutions culture environment for prostate epithelial cells derived from normal human prostate (eg, Primary Prostate Epithelial Cells, Normal, Human, ATCC PCS-440-010). The serum-free medium formulation is designed to support normal prostate cell morphology as well as promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-3922 Supplier ATCC Supplier No. PCS440040
Prostate Epithelial Cell Growth Kit (ATCC PCS-440-040) contains components that when added to Prostate Epithelial Cell Basal Medium (ATCC PCS-440-030) create a complete ATCC Primary Cell Solutions culture environment for prostate epithelial cells derived from normal human prostate (eg, Primary Prostate Epithelial Cells, Normal, Human, ATCC PCS-440-010). The serum-free medium formulation is designed to support normal prostate cell morphology as well as promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.