Learn More
Lonza Walkersville PrEC – Human Prostate Epithelial Cells
Supplier: Lonza Walkersville CC2555
Prostate cells provide a glandular function in the body by generating fluid which serves several functions in reproduction. Lonza's Human Prostate Cells are available from epithelial, smooth muscle, and stromal tissue. Each cell type has an optimized growth media for best results in culture.Prostate Epithelial Cells (PrEC) are guaranteed through 15 population doublings and stain positive for cytokeratin (clone 8.13). Cryopreserved PrEC are shipped in first or second passage.Benefits:Guaranteed through 15 population doublings Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate ofAnalysis is provided for each cell lot purchased Applications:Benign prostatic hyperplasia (BPH)ProstitisProstate cancerProcreation researchDrug discoveryMale prostate physiologyContent:1 x Cryopreserved ampule PrEC containing ≥500,000 cells