Learn More
Cayman Chemical ProstatIc ACD Phosphatsem
SDP
Catalog No. 502916601
Encompass_Preferred
Source: Recombinant mouse C-terminal His-tagged prostatic acid phosphatase expressed in HEK293 cells • Amino acids: 32-381 • MW: 42 kDa
Catalog No. 50-291-6601 Supplier Cayman Chemical Supplier No. 38071100 UG
Source: Recombinant mouse C-terminal His-tagged prostatic acid phosphatase expressed in HEK293 cells • Amino acids: 32-381 • MW: 42 kDa