Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prostatic Acid Phosphatase Antibody (YA5619) | Predicted band size: 58 kDa; Observed band size: 68 kDa | 50 UL
SDP

Catalog No. 5000446582
Encompass_Preferred

The Prostatic Acid Phosphatase (PAP) Antibody targets PAP, a non-specific tyrosine phosphatase critical for dephosphorylating various substrates under acidic conditions. PAP plays a role in prostate cancer as a tumor suppressor and is involved in pain suppression in dorsal root ganglion neurons. It also forms amyloid fibrils that can enhance viral infections such as HIV.

  • Host: Mouse
  • Reactivity: Human, Mouse, Rat
  • Conjugation: Non-conjugated
  • Formulation: Supplied in PBS, 50% glycerol, 0.05% Proclin 300, 0.05% BSA
  • Appearance: Liquid
  • Applications: IHC-P (1:200-1000), WB (1:500-2000), ICC/IF (1:100-500), ELISA (1:1000-5000)
  • Storage conditions: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Catalog No. 50-004-46582 Supplier Medchemexpress LLC Supplier No. HYP8592750UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Prostatic Acid Phosphatase (PAP) Antibody targets PAP, a non-specific tyrosine phosphatase critical for dephosphorylating various substrates under acidic conditions. PAP plays a role in prostate cancer as a tumor suppressor and is involved in pain suppression in dorsal root ganglion neurons. It also forms amyloid fibrils that can enhance viral infections such as HIV.

  • Host: Mouse
  • Reactivity: Human, Mouse, Rat
  • Conjugation: Non-conjugated
  • Formulation: Supplied in PBS, 50% glycerol, 0.05% Proclin 300, 0.05% BSA
  • Appearance: Liquid
  • Applications: IHC-P (1:200-1000), WB (1:500-2000), ICC/IF (1:100-500), ELISA (1:1000-5000)
  • Storage conditions: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.