Learn More
AdipoGen Prosulfocarb sulfoxide
SDP
Catalog No. 502560182
Encompass_Preferred
Chemical. CAS 51954-81-5. Formula C14H21NO2S. MW 267.39. Metabolite of the herbicide Prosulfocarb. Can be used as a reference compound.
Catalog No. 50-256-0182 Supplier AdipoGen Supplier No. CDXP0507M050
Chemical. CAS 51954-81-5. Formula C14H21NO2S. MW 267.39. Metabolite of the herbicide Prosulfocarb. Can be used as a reference compound.