Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ ProSurface Spray

Catalog No. 19910706 Shop All Medicom Products
Change view
Click to view available options
Volume (English):
32 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-910-706 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-910-706 Supplier Medicom™ Supplier No. 40066

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

ProSurface disinfectant provides powerful one-step cleaning with a best-in-class contact time of just one minute. ProSurface eliminates 48 pathogens, including bacteria, viruses and fungi, without damaging surfaces.

  • Proprietary TotalClean™ technology provides powerful 1-step cleaning and disinfection in a best-in-class contact time: just 1 minute
  • ProSurface kills more pathogens than most competitors - eliminates 48 pathogens, including bacteria, viruses and fungi
  • Surface-safe on most non-porous surfaces
  • No residue (low quat content)
  • Fragrance-free
  • Low toxicity - no gloves or protective eyewear required for handling
  • EPA registered - i.e., all claims have been validated
  • Treated surface must remain visibly wet for one minute

Specifications

Sterility Non-sterile
Composition Alcohol
Container Type Spray
Disposable Single-use
For Use With (Application) Surfaces
Volume (English) 32 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.