Learn More
Medicom™ ProSurface Wipes
Catalog No. 19910704 Shop All Medicom Products
Change view
Click to view available options
Dimensions (L x W):
6 x 6.75 in.
Catalog No. Dimensions (L x W)
19-910-704 6 x 6.75 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-910-704 Supplier Medicom™ Supplier No. 40060
This item is not returnable. View return policy
ProSurface disinfectant provides powerful one-step cleaning with a best-in-class contact time of just one minute. ProSurface eliminates 48 pathogens, including bacteria, viruses and fungi, without damaging surfaces.
  • Proprietary TotalClean™ technology provides powerful 1-step cleaning and disinfection in a best-in-class contact time: just 1 minute
  • ProSurface kills more pathogens than most competitors - eliminates 48 pathogens, including bacteria, viruses and fungi
  • Surface-safe on most non-porous surfaces
  • No residue (low quat content)
  • Fragrance-free
  • Low toxicity - no gloves or protective eyewear required for handling
  • EPA registered - i.e., all claims have been validated
  • Treated surface must remain visibly wet for one minute
Specifications
Product Type Wipes
Material Polypropylene
Autoclavable Not Autoclavable
Disposable Single-use
Dimensions (L x W) 6 x 6.75 in.
For Use With (Application) Surfaces
Sterility Non-sterile