Learn More
New England Biolabs, Inc. Protein A Magnetic Beads 1 ml
SDP
Catalog No. 50812661
Encompass
Protein A Magnetic Beads are an affinity matrix for the small-scale isolation and purification of immunoglobulins. Specifically, the beads consist of a truncated form of recombinant Protein A that is covalently coupled to a nonporous superparamagnetic particle. The protein is coupled through a linkage that is stable and leak resistant over a wide pH range.
Catalog No. 50-812-661 Supplier New England Biolabs, Inc. Supplier No. S1425S
Protein A Magnetic Beads are an affinity matrix for the small-scale isolation and purification of immunoglobulins. Specifically, the beads consist of a truncated form of recombinant Protein A that is covalently coupled to a nonporous superparamagnetic particle. The protein is coupled through a linkage that is stable and leak resistant over a wide pH range.