Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protac Axl degrader 2 | 00-00-0 | 98.0% | 713.79 g/mol | C38H39N11O4 | 5 MG
SDP

Catalog No. 5000221216
Encompass_Preferred

PROTAC Axl Degrader 2 is a proteolysis targeting chimera designed to induce selective degradation of the Axl receptor tyrosine kinase for research applications. It is intended for in vitro studies of Axl biology and PROTAC-mediated protein degradation.

  • Proteolysis targeting chimera (PROTAC) that induces selective degradation of the Axl receptor.
  • Targets TAM family receptor tyrosine kinases via a PROTAC mechanism.
  • Molecular formula C38H39N11O4 and molecular weight 713.79 g/mol.
  • Purity approximately 98.0% as supplied.
  • Recommended storage: protect from light and store under nitrogen; in solvent, -80°C for long-term, -20°C short-term.
  • For research use only; not validated for clinical or therapeutic use.

Catalog No. 50-002-21216 Supplier Medchemexpress LLC Supplier No. HY1446275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC Axl Degrader 2 is a proteolysis targeting chimera designed to induce selective degradation of the Axl receptor tyrosine kinase for research applications. It is intended for in vitro studies of Axl biology and PROTAC-mediated protein degradation.

  • Proteolysis targeting chimera (PROTAC) that induces selective degradation of the Axl receptor.
  • Targets TAM family receptor tyrosine kinases via a PROTAC mechanism.
  • Molecular formula C38H39N11O4 and molecular weight 713.79 g/mol.
  • Purity approximately 98.0% as supplied.
  • Recommended storage: protect from light and store under nitrogen; in solvent, -80°C for long-term, -20°C short-term.
  • For research use only; not validated for clinical or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.