Learn More
Medchemexpress LLC PROTAC Axl Degrader 2 | 3033401-45-2 | 98.01% | C38H39N11O4 | 1 MG
SDP
Catalog No. 5000356287
Encompass_Preferred
PROTAC Axl Degrader 2 is a potent and selective PROTAC Axl degrader with an IC50 of 1.61 μM. It exhibits anti-proliferation activity and anti-migration activity in vitro. This compound also induces methuosis.
  • Potent and selective PROTAC Axl degrader.
  • Shows anti-proliferation activity in vitro.
  • Demonstrates anti-migration activity in vitro.
  • Induces methuosis.
  • Decreases the abundance of AXL in MDA-MB-231 cells.
  • Significantly inhibits migration in MDA-MB-231 and 4T1 cells.
  • Does not induce systemic toxicity at 25 mg/kg (i.p.) in female balb/c nude mice.
Catalog No. 50-003-56287 Supplier Medchemexpress LLC Supplier No. HY1446271MG
PROTAC Axl Degrader 2 is a potent and selective PROTAC Axl degrader with an IC50 of 1.61 μM. It exhibits anti-proliferation activity and anti-migration activity in vitro. This compound also induces methuosis.
  • Potent and selective PROTAC Axl degrader.
  • Shows anti-proliferation activity in vitro.
  • Demonstrates anti-migration activity in vitro.
  • Induces methuosis.
  • Decreases the abundance of AXL in MDA-MB-231 cells.
  • Significantly inhibits migration in MDA-MB-231 and 4T1 cells.
  • Does not induce systemic toxicity at 25 mg/kg (i.p.) in female balb/c nude mice.