Learn More
Medchemexpress LLC Protac Bcl-xL ligand-1 | 3029365-95-2 | 99.1% | C32H29IN4O4S2 | 10MG
SDP
Catalog No. 5000259791
Encompass_Preferred
PROTAC Bcl-xL ligand-1 is a small-molecule ligand for the anti-apoptotic protein Bcl-xL, intended as a building block for proteolysis-targeting chimera (PROTAC) synthesis and related biochemical research. The compound is characterized by CAS 3029365-95-2, molecular formula C32H29IN4O4S2, molecular weight 724.63, and a manufacturer-reported purity of 99.06%. Safety data and handling information are provided in the product SDS.
  • High reported purity (99.06%).
  • Characterized by CAS number and molecular identifiers.
  • Suitable for use in PROTAC synthesis and ligand development.
  • Available in multiple small-molecule quantities for research use.
  • Safety data sheet and handling guidance available.
Catalog No. 50-002-59791 Supplier Medchemexpress LLC Supplier No. HY13930410MG
PROTAC Bcl-xL ligand-1 is a small-molecule ligand for the anti-apoptotic protein Bcl-xL, intended as a building block for proteolysis-targeting chimera (PROTAC) synthesis and related biochemical research. The compound is characterized by CAS 3029365-95-2, molecular formula C32H29IN4O4S2, molecular weight 724.63, and a manufacturer-reported purity of 99.06%. Safety data and handling information are provided in the product SDS.
  • High reported purity (99.06%).
  • Characterized by CAS number and molecular identifiers.
  • Suitable for use in PROTAC synthesis and ligand development.
  • Available in multiple small-molecule quantities for research use.
  • Safety data sheet and handling guidance available.