Learn More
Medchemexpress LLC PROTAC BCR-ABL Degrader-1 | 3079412-59-9 | C43H40N10O6 | 1 MG
Supplier: Medchemexpress LLC HY1534151MG
PROTAC BCR-ABL Degrader-1 is a PROTAC with a 2-oxoethyl linker. It induces Bcr-Abl degradation in a ubiquitin-proteasome-dependent manner, exhibiting antiproliferative activity against K562 cells. This product is for research use only and has the potential for cancer research.
- Induces Bcr-Abl degradation
- Exhibits antiproliferative activity against K562 cells
- Potential for cancer research
- Acts as an inhibitor