Learn More
Medchemexpress LLC PROTAC BCR-ABL Degrader-1 | 3079412-59-9 | C43H40N10O6 | 100 MG
SDP
Catalog No. 5000372648
Encompass_Preferred
PROTAC BCR-ABL Degrader-1 is a PROTAC with a 2-oxoethyl linker. It functions by inducing Bcr-Abl degradation through a ubiquitin-proteasome-dependent mechanism. This compound demonstrates antiproliferative activity against K562 cells, indicating its potential for use in cancer research.
  • Induces Bcr-Abl degradation
  • Functions in a ubiquitin-proteasome-dependent manner
  • Exhibits antiproliferative activity against K562 cells
  • Potential for cancer research
  • Features a 2-oxoethyl linker
Catalog No. 50-003-72648 Supplier Medchemexpress LLC Supplier No. HY153415100MG
PROTAC BCR-ABL Degrader-1 is a PROTAC with a 2-oxoethyl linker. It functions by inducing Bcr-Abl degradation through a ubiquitin-proteasome-dependent mechanism. This compound demonstrates antiproliferative activity against K562 cells, indicating its potential for use in cancer research.
  • Induces Bcr-Abl degradation
  • Functions in a ubiquitin-proteasome-dependent manner
  • Exhibits antiproliferative activity against K562 cells
  • Potential for cancer research
  • Features a 2-oxoethyl linker