Learn More
Medchemexpress LLC PROTAC BCR-ABL Degrader-1 | 3079412-59-9 | C43H40N10O6 | 5 MG
SDP
Catalog No. 5000373533
Encompass_Preferred
PROTAC BCR-ABL Degrader-1, also known as compound PROTAC 1, is a PROTAC (Proteolysis Targeting Chimera) featuring a 2-oxoethyl linker. This compound functions by inducing Bcr-Abl degradation through a ubiquitin-proteasome-dependent mechanism. It has demonstrated antiproliferative activity against K562 cells, indicating its potential for use in cancer research.
  • Utilizes a PROTAC approach with a 2-oxoethyl linker.
  • Induces Bcr-Abl degradation.
  • Operates via a ubiquitin-proteasome-dependent manner.
  • Exhibits antiproliferative activity against K562 cells.
  • Has the potential for studying cancer.
Catalog No. 50-003-73533 Supplier Medchemexpress LLC Supplier No. HY1534155MG
PROTAC BCR-ABL Degrader-1, also known as compound PROTAC 1, is a PROTAC (Proteolysis Targeting Chimera) featuring a 2-oxoethyl linker. This compound functions by inducing Bcr-Abl degradation through a ubiquitin-proteasome-dependent mechanism. It has demonstrated antiproliferative activity against K562 cells, indicating its potential for use in cancer research.
  • Utilizes a PROTAC approach with a 2-oxoethyl linker.
  • Induces Bcr-Abl degradation.
  • Operates via a ubiquitin-proteasome-dependent manner.
  • Exhibits antiproliferative activity against K562 cells.
  • Has the potential for studying cancer.