Learn More
Medchemexpress LLC PROTAC BCR-ABL1 ligand 1 | 2489876-34-6 | 99.17% | 50 MG
SDP
Catalog No. 5000341061
Encompass_Preferred
PROTAC BCR-ABL1 ligand 1, also known as compound GMB-475, is a PROTAC ligand that allosterically targets the BCR-ABL1 protein. It functions by recruiting the E3 ligase Von Hippel-Lindau, which leads to the ubiquitination and subsequent degradation of BCR-ABL1.
  • Allosterically targets the BCR-ABL1 protein.
  • Recruits the E3 ligase Von Hippel-Lindau.
  • Induces ubiquitination and subsequent degradation of BCR-ABL1.
  • Inhibits cell proliferation with IC50 values of 1.11 μM for WT cells, 1.98 μM for T315I cells, and 0.37 μM for G250E cells.
  • Induces the degradation of c-ABL1.
  • Inhibits downstream signaling through the STAT5 pathway in K562 and Ba/F3 cells.
Catalog No. 50-003-41061 Supplier Medchemexpress LLC Supplier No. HY13029750MG
PROTAC BCR-ABL1 ligand 1, also known as compound GMB-475, is a PROTAC ligand that allosterically targets the BCR-ABL1 protein. It functions by recruiting the E3 ligase Von Hippel-Lindau, which leads to the ubiquitination and subsequent degradation of BCR-ABL1.
  • Allosterically targets the BCR-ABL1 protein.
  • Recruits the E3 ligase Von Hippel-Lindau.
  • Induces ubiquitination and subsequent degradation of BCR-ABL1.
  • Inhibits cell proliferation with IC50 values of 1.11 μM for WT cells, 1.98 μM for T315I cells, and 0.37 μM for G250E cells.
  • Induces the degradation of c-ABL1.
  • Inhibits downstream signaling through the STAT5 pathway in K562 and Ba/F3 cells.