Learn More
Medchemexpress LLC PROTAC BCR-ABL1 ligand 1 | 2489876-34-6 | 99.2% | 10 MM * 1 ML
SDP
Catalog No. 5000338654
Encompass_Preferred
PROTAC BCR-ABL1 ligand 1, also known as compound GMB-475, is a PROTAC ligand that allosterically targets the BCR-ABL1 protein. It recruits the E3 ligase Von Hippel-Lindau, leading to the ubiquitination and subsequent degradation of BCR-ABL1.
  • Exhibits cell proliferation with specific IC50 values for BCR-ABL1 WT, T315I, and G250E cells.
  • Induces the degradation of BCR-ABL1 and c-ABL1.
  • Inhibits downstream signaling via the STAT5 pathway.
  • Demonstrates dose- and time-dependent action in K562 and Ba/F3 cells.
Catalog No. 50-003-38654 Supplier Medchemexpress LLC Supplier No. HY13029710MM/1ML
PROTAC BCR-ABL1 ligand 1, also known as compound GMB-475, is a PROTAC ligand that allosterically targets the BCR-ABL1 protein. It recruits the E3 ligase Von Hippel-Lindau, leading to the ubiquitination and subsequent degradation of BCR-ABL1.
  • Exhibits cell proliferation with specific IC50 values for BCR-ABL1 WT, T315I, and G250E cells.
  • Induces the degradation of BCR-ABL1 and c-ABL1.
  • Inhibits downstream signaling via the STAT5 pathway.
  • Demonstrates dose- and time-dependent action in K562 and Ba/F3 cells.