Learn More
Medchemexpress LLC Protac BCR-ABL1 ligand 1 | 2489876-34-6 | 99.2% | 347.29 g/mol | C17H12F3N3O2 | 5 MG
SDP
Catalog No. 5000263163
Encompass_Preferred
PROTAC BCR-ABL1 ligand 1 is a small-molecule PROTAC ligand (GMB-475) that allosterically binds BCR-ABL1 and recruits the Von Hippel-Lindau (VHL) E3 ligase to promote ubiquitination and proteasomal degradation. Supplied for research use as a solid, it is provided at high purity for biochemical and cell-based studies.
  • Allosteric ligand for BCR-ABL1 that promotes target degradation.
  • Recruits VHL E3 ligase to facilitate ubiquitination.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight 347.29 g/mol, compatible with screening workflows.
  • Available in small research quantities for screening and validation.
Catalog No. 50-002-63163 Supplier Medchemexpress LLC Supplier No. HY1302975MG
PROTAC BCR-ABL1 ligand 1 is a small-molecule PROTAC ligand (GMB-475) that allosterically binds BCR-ABL1 and recruits the Von Hippel-Lindau (VHL) E3 ligase to promote ubiquitination and proteasomal degradation. Supplied for research use as a solid, it is provided at high purity for biochemical and cell-based studies.
  • Allosteric ligand for BCR-ABL1 that promotes target degradation.
  • Recruits VHL E3 ligase to facilitate ubiquitination.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight 347.29 g/mol, compatible with screening workflows.
  • Available in small research quantities for screening and validation.