Learn More
Medchemexpress LLC HY-43723 5mg Medchemexpress, PROTAC BET-binding moiety 2 CAS:916493-82-8 Purity:>98%
SDP
Catalog No. 501932864
Encompass_Preferred
Medchemexpress, HY-43723 5mg PROTAC BET-binding moiety 2 CAS:916493-82-8 PROTAC BET-binding moiety 2 is an inhibitor of BET bromodomain [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2864 Supplier Medchemexpress LLC Supplier No. HY437235MG
Medchemexpress, HY-43723 5mg PROTAC BET-binding moiety 2 CAS:916493-82-8 PROTAC BET-binding moiety 2 is an inhibitor of BET bromodomain [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.