Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PROTAC BET-binding moiety 1 | 2093387-77-8 | 80.4% | 487.51 | C25H25N7O4 | 1 ML
SDP

Catalog No. 5000240583
Encompass_Preferred

PROTAC BET-binding moiety 1 is a small-molecule intermediate used in the synthesis of high-affinity BET (bromodomain and extra-terminal) inhibitors. Supplied as a ready-to-use DMSO solution (10 mM, 1 mL), it is intended for medicinal chemistry and PROTAC development targeting BRD2, BRD3, BRD4, and BRDT.

  • Ready-to-use 10 mM solution in DMSO.
  • Suitable for synthesis of BET inhibitors and PROTACs.
  • Molecular formula C25H25N7O4 and molecular weight 487.51.
  • Reported purity 80.4%.
  • CAS number 2093387-77-8 for unambiguous identification.

Catalog No. 50-002-40583 Supplier Medchemexpress LLC Supplier No. HY10745110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC BET-binding moiety 1 is a small-molecule intermediate used in the synthesis of high-affinity BET (bromodomain and extra-terminal) inhibitors. Supplied as a ready-to-use DMSO solution (10 mM, 1 mL), it is intended for medicinal chemistry and PROTAC development targeting BRD2, BRD3, BRD4, and BRDT.

  • Ready-to-use 10 mM solution in DMSO.
  • Suitable for synthesis of BET inhibitors and PROTACs.
  • Molecular formula C25H25N7O4 and molecular weight 487.51.
  • Reported purity 80.4%.
  • CAS number 2093387-77-8 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.