Learn More
Medchemexpress LLC Protac BET-binding moiety 2 | 916493-82-8 | 99.8% | 10 MG
SDP
Catalog No. 5000297898
Encompass_Preferred
Small-molecule BET bromodomain inhibitor intended as the BET-binding ligand component in PROTAC constructs and for biochemical and cell-based research applications. Supplied as a solid or as a 10 mM solution in DMSO, with reported high purity and identifying registry information for compound verification.
  • Functions as a BET bromodomain ligand for PROTAC design and target engagement studies.
  • Available as solid powder and as a 10 mM solution in DMSO for assay flexibility.
  • High reported purity suitable for research use.
  • Characterized by CAS 916493-82-8 and molecular weight 444.89 for identification.
  • Provided for research use only; not for human or clinical use.
Catalog No. 50-002-97898 Supplier Medchemexpress LLC Supplier No. HY4372310MG
Small-molecule BET bromodomain inhibitor intended as the BET-binding ligand component in PROTAC constructs and for biochemical and cell-based research applications. Supplied as a solid or as a 10 mM solution in DMSO, with reported high purity and identifying registry information for compound verification.
  • Functions as a BET bromodomain ligand for PROTAC design and target engagement studies.
  • Available as solid powder and as a 10 mM solution in DMSO for assay flexibility.
  • High reported purity suitable for research use.
  • Characterized by CAS 916493-82-8 and molecular weight 444.89 for identification.
  • Provided for research use only; not for human or clinical use.