Learn More
Medchemexpress LLC HY-103633 10mg Medchemexpress, PROTAC BET Degrader-1 CAS:2093386-22-0 Purity:>98%
SDP
Catalog No. 501968145
Encompass_Preferred
Medchemexpress, HY-103633 10mg PROTAC BET Degrader-1 CAS:2093386-22-0 PROTAC BET Degrader-1 is a potent BET degrader based on PROTAC, decreasing BRD2, BRD3, and BRD4 protein levels at low concentration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8145 Supplier Medchemexpress LLC Supplier No. HY10363310MG
Medchemexpress, HY-103633 10mg PROTAC BET Degrader-1 CAS:2093386-22-0 PROTAC BET Degrader-1 is a potent BET degrader based on PROTAC, decreasing BRD2, BRD3, and BRD4 protein levels at low concentration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.