Learn More
Medchemexpress LLC PROTAC BET degrader-1 | 2093386-22-0 | 99.4% | 871.9 g/mol | C44H45N11O9 | 100 MG
SDP
Catalog No. 5000246830
Encompass_Preferred
PROTAC BET Degrader-1 is a proteolysis-targeting chimera (PROTAC) that links Cereblon and BET ligands to promote ubiquitin-mediated degradation of BET family proteins BRD2, BRD3, and BRD4 at low concentrations. It is supplied for research use in studies of BET protein biology and targeted protein degradation.
  • Induces degradation of BRD2, BRD3, and BRD4 at low nanomolar concentrations.
  • High purity (99.4%) suitable for biochemical and cell-based assays.
  • Available as solid or as DMSO solution for experimental flexibility.
  • High solubility in DMSO (≥ 50 mg/mL) for convenient dosing.
  • Stable when stored per manufacturer recommendations for long-term experiments.
Catalog No. 50-002-46830 Supplier Medchemexpress LLC Supplier No. HY103633100MG
PROTAC BET Degrader-1 is a proteolysis-targeting chimera (PROTAC) that links Cereblon and BET ligands to promote ubiquitin-mediated degradation of BET family proteins BRD2, BRD3, and BRD4 at low concentrations. It is supplied for research use in studies of BET protein biology and targeted protein degradation.
  • Induces degradation of BRD2, BRD3, and BRD4 at low nanomolar concentrations.
  • High purity (99.4%) suitable for biochemical and cell-based assays.
  • Available as solid or as DMSO solution for experimental flexibility.
  • High solubility in DMSO (≥ 50 mg/mL) for convenient dosing.
  • Stable when stored per manufacturer recommendations for long-term experiments.