Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protac BET degrader-1 | 2093386-22-0 | 99.4% | 871.90 g/mol | C44H45N11O9 | 1 ML
SDP

Catalog No. 5000246831
Encompass_Preferred

PROTAC BET Degrader-1 is a proteolysis-targeting chimera that links a Cereblon ligand to a BET-family ligand to promote ubiquitination and proteasomal degradation of BET proteins BRD2, BRD3, and BRD4. It is used in biochemical and cellular assays to investigate BET protein degradation and downstream epigenetic effects.

  • Induces selective degradation of BRD2, BRD3, and BRD4 at low nanomolar concentrations.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.4% by LCMS).
  • Molecular formula C44H45N11O9; molecular weight 871.90 g/mol.
  • Storage: powder -20°C for long term; in solvent -80°C recommended.

Catalog No. 50-002-46831 Supplier Medchemexpress LLC Supplier No. HY10363310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC BET Degrader-1 is a proteolysis-targeting chimera that links a Cereblon ligand to a BET-family ligand to promote ubiquitination and proteasomal degradation of BET proteins BRD2, BRD3, and BRD4. It is used in biochemical and cellular assays to investigate BET protein degradation and downstream epigenetic effects.

  • Induces selective degradation of BRD2, BRD3, and BRD4 at low nanomolar concentrations.
  • Provided as a 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.4% by LCMS).
  • Molecular formula C44H45N11O9; molecular weight 871.90 g/mol.
  • Storage: powder -20°C for long term; in solvent -80°C recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.