Learn More
Medchemexpress LLC PROTAC BET Degrader-10 | 1957234-97-7 | 99.4% | 783.29 | 50 MG
SDP
Catalog No. 5000435799
Encompass_Preferred
PROTAC BET Degrader-10 (dBET37) is a potent BET protein BRD4 degrader. It is connected by ligands for Cereblon and BRD4, and has a DC50 of 49 nM. This product is for research use only and not sold to patients. PROTACs use two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Potent BET protein BRD4 degrader
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Connected by ligands for Cereblon and BRD4
  • DC50 of 49 nM
Catalog No. 50-004-35799 Supplier Medchemexpress LLC Supplier No. HY11271850MG
PROTAC BET Degrader-10 (dBET37) is a potent BET protein BRD4 degrader. It is connected by ligands for Cereblon and BRD4, and has a DC50 of 49 nM. This product is for research use only and not sold to patients. PROTACs use two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Potent BET protein BRD4 degrader
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Connected by ligands for Cereblon and BRD4
  • DC50 of 49 nM