Learn More
Medchemexpress LLC Protac BET degrader-10 | 1957234-97-7 | 99.4% | 783.29 g/mol | C39H39ClN8O6S | 5 MG
SDP
Catalog No. 5000262893
Encompass_Preferred
PROTAC BET Degrader-10 is a small-molecule proteolysis-targeting chimera designed to induce degradation of BET family proteins, primarily BRD4. Reported in patent WO2017007612A1 (example 37), the compound links cereblon and BRD4 ligands and has a reported DC50 of 49 nM. Typical analytical data include CAS 1957234-97-7, molecular formula C39H39ClN8O6S, molecular weight 783.29 g/mol, and purity ~99.39%.
  • Induces selective BRD4 degradation in cellular assays.
  • Reported DC50 of approximately 49 nM for BRD4 degradation.
  • Available as solid and as a 10 mM solution in DMSO for assay flexibility.
  • High chemical purity suitable for biochemical and cellular studies.
  • Applicable to PROTAC mechanism-of-action and epigenetic research.
Catalog No. 50-002-62893 Supplier Medchemexpress LLC Supplier No. HY1127185MG
PROTAC BET Degrader-10 is a small-molecule proteolysis-targeting chimera designed to induce degradation of BET family proteins, primarily BRD4. Reported in patent WO2017007612A1 (example 37), the compound links cereblon and BRD4 ligands and has a reported DC50 of 49 nM. Typical analytical data include CAS 1957234-97-7, molecular formula C39H39ClN8O6S, molecular weight 783.29 g/mol, and purity ~99.39%.
  • Induces selective BRD4 degradation in cellular assays.
  • Reported DC50 of approximately 49 nM for BRD4 degradation.
  • Available as solid and as a 10 mM solution in DMSO for assay flexibility.
  • High chemical purity suitable for biochemical and cellular studies.
  • Applicable to PROTAC mechanism-of-action and epigenetic research.