Learn More
Medchemexpress LLC HY-114228 10mg Medchemexpress, PROTAC BET degrader-2 CAS:2093388-33-9 Purity:>98%
SDP
Catalog No. 501969307
Encompass_Preferred
Medchemexpress, HY-114228 10mg PROTAC BET degrader-2 CAS:2093388-33-9 PROTAC BET degrader-2 is a highly potent degrader of Bromodomain and Extra-Terminal (BET) proteins with an IC50 value of 9.6 nM in cell growth inhibition in the RS4;11 cells and capable of achieving tumor regression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9307 Supplier Medchemexpress LLC Supplier No. HY11422810MG
Medchemexpress, HY-114228 10mg PROTAC BET degrader-2 CAS:2093388-33-9 PROTAC BET degrader-2 is a highly potent degrader of Bromodomain and Extra-Terminal (BET) proteins with an IC50 value of 9.6 nM in cell growth inhibition in the RS4;11 cells and capable of achieving tumor regression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.