Learn More
Medchemexpress LLC PROTAC BET degrader- 25mg | 3031993-61-7 | 25 MG
SDP
Catalog No. 5000228835
Encompass_Preferred
PROTAC BET Degrader-3 is a proteolysis-targeting chimera that links a von Hippel-Lindau (VHL) E3 ligase ligand to a BET-family ligand to induce targeted BET protein degradation. It is supplied as a light yellow solid for research use only.
  • Proteolysis-targeting chimera linking VHL and BET ligands.
  • Designed to induce targeted degradation of BET-family proteins in biochemical and cellular assays.
  • High purity suitable for research-grade experiments.
  • Available in milligram-scale quantities for early-stage research.
  • Stable as a powder at -20°C; in solution, store at -80°C for extended stability.
Catalog No. 50-002-28835 Supplier Medchemexpress LLC Supplier No. HY11422925MG
PROTAC BET Degrader-3 is a proteolysis-targeting chimera that links a von Hippel-Lindau (VHL) E3 ligase ligand to a BET-family ligand to induce targeted BET protein degradation. It is supplied as a light yellow solid for research use only.
  • Proteolysis-targeting chimera linking VHL and BET ligands.
  • Designed to induce targeted degradation of BET-family proteins in biochemical and cellular assays.
  • High purity suitable for research-grade experiments.
  • Available in milligram-scale quantities for early-stage research.
  • Stable as a powder at -20°C; in solution, store at -80°C for extended stability.