Learn More
Medchemexpress LLC HY-129938 1mg Medchemexpress, PROTAC BRD4 degrader for PAC-1 CAS:2417370-39-7 Purity:>98%
SDP
Catalog No. 502030180
Encompass_Preferred
Medchemexpress, HY-129938 1mg PROTAC BRD4 degrader for PAC-1 CAS:2417370-39-7 PROTAC BRD4 degrader for PAC-1 (compound 5), a PROTAC-linker Conjugate for PAC, comprises the chimeric BET degrader GNE-987 and disulfide-containing linker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0180 Supplier Medchemexpress LLC Supplier No. HY1299381MG
Medchemexpress, HY-129938 1mg PROTAC BRD4 degrader for PAC-1 CAS:2417370-39-7 PROTAC BRD4 degrader for PAC-1 (compound 5), a PROTAC-linker Conjugate for PAC, comprises the chimeric BET degrader GNE-987 and disulfide-containing linker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.