Learn More
TARGETMOL CHEMICALS INC PROTAC BRD4 LIGAND-1 5MG
SDP
Catalog No. 502581441
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. PROTAC BRD4 ligand-1 is a ligand for BRD4 for Protac GNE-987 and an inhibitor of BET. purity: 99%
Catalog No. 50-258-1441 Supplier TARGETMOL CHEMICALS INC Supplier No. T125515MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. PROTAC BRD4 ligand-1 is a ligand for BRD4 for Protac GNE-987 and an inhibitor of BET. purity: 99%