Learn More
Medchemexpress LLC HY-107445A 5mg Medchemexpress, PROTAC BRD9-binding moiety 1 hydrochloride CAS: Purity:>98%
SDP
Catalog No. 501932285
Encompass_Preferred
Medchemexpress, HY-107445A 5mg PROTAC BRD9-binding moiety 1 hydrochloride CAS: PROTAC BRD9-binding moiety 1 hydrochloride is a compound that binds to BRD9 , and used for inhibiting BRD9 activity, based on PROTAC . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2285 Supplier Medchemexpress LLC Supplier No. HY107445A5MG
Medchemexpress, HY-107445A 5mg PROTAC BRD9-binding moiety 1 hydrochloride CAS: PROTAC BRD9-binding moiety 1 hydrochloride is a compound that binds to BRD9 , and used for inhibiting BRD9 activity, based on PROTAC . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.