Learn More
Medchemexpress LLC HY-103632 10mg Medchemexpress, PROTAC BRD9 Degrader-1 CAS:2097971-01-0 Purity:>98%
SDP
Catalog No. 501968144
Encompass_Preferred
Medchemexpress, HY-103632 10mg PROTAC BRD9 Degrader-1 CAS:2097971-01-0 PROTAC BRD9 Degrader-1 is a lead PROTAC BRD9 chemical degrader (IC50=13.5 nM), which can be used as a selective probe useful for the study of BAF complex biology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8144 Supplier Medchemexpress LLC Supplier No. HY10363210MG
Medchemexpress, HY-103632 10mg PROTAC BRD9 Degrader-1 CAS:2097971-01-0 PROTAC BRD9 Degrader-1 is a lead PROTAC BRD9 chemical degrader (IC50=13.5 nM), which can be used as a selective probe useful for the study of BAF complex biology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.