Learn More
Medchemexpress LLC PROTAC BTK Degrader-10 | 2988804-46-0 | 771.91 | 5 MG
SDP
Catalog No. 5000394355
Encompass_Preferred
PROTAC BTK Degrader-10 is a PROTAC degrader for BTK. It can be used for research of chronic lymphocytic leukemia (CLL). It consists of a pink ligand for target protein (HY-170758), a black linker (HY-W392857), and a blue ligand for E3 ligase Cereblon (HY-W733176).
  • PROTAC degrader for BTK
  • Applicable for research on chronic lymphocytic leukemia (CLL)
Catalog No. 50-003-94355 Supplier Medchemexpress LLC Supplier No. HY1609665MG
PROTAC BTK Degrader-10 is a PROTAC degrader for BTK. It can be used for research of chronic lymphocytic leukemia (CLL). It consists of a pink ligand for target protein (HY-170758), a black linker (HY-W392857), and a blue ligand for E3 ligase Cereblon (HY-W733176).
  • PROTAC degrader for BTK
  • Applicable for research on chronic lymphocytic leukemia (CLL)