Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protac CBP/p300 degrader-1 | 2484739-48-0 | 98.3% | 893.98 | C46H53F2N11O6 | 5 MG
SDP

Catalog No. 5000222383
Encompass_Preferred

PROTAC CBP/P300 Degrader-1 is a small-molecule proteolysis targeting chimera designed to induce selective degradation of the epigenetic coactivators CBP and p300. It demonstrates potent degradation activity and reduces viability in multiple cancer cell lines, and is intended for research use in cellular and biochemical assays.

  • Targets and degrades CBP and p300 in cell-based assays.
  • Potently inhibits viability of multiple cancer cell lines.
  • Molecular formula C46H53F2N11O6.
  • Molecular weight 893.98 Da.
  • Supplied in milligram quantities for research applications.

Catalog No. 50-002-22383 Supplier Medchemexpress LLC Supplier No. HY1385365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC CBP/P300 Degrader-1 is a small-molecule proteolysis targeting chimera designed to induce selective degradation of the epigenetic coactivators CBP and p300. It demonstrates potent degradation activity and reduces viability in multiple cancer cell lines, and is intended for research use in cellular and biochemical assays.

  • Targets and degrades CBP and p300 in cell-based assays.
  • Potently inhibits viability of multiple cancer cell lines.
  • Molecular formula C46H53F2N11O6.
  • Molecular weight 893.98 Da.
  • Supplied in milligram quantities for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.