Learn More
Cayman Chemical PROTAC CRBN Degrader-1, Purity: ≥95%, CAS: 2358775-70-7, 1 mg
SDP
Catalog No. 502945490
Encompass_Preferred
A PROTAC that drives CRBN degradation (DC50 = 200 nM in HeLa cells)
Catalog No. 50-294-5490 Supplier Cayman Chemical Supplier No. 418781 MG
A PROTAC that drives CRBN degradation (DC50 = 200 nM in HeLa cells)