Learn More
Medchemexpress LLC PROTAC CRBN Degrader-1 | 2358775-70-7 | 98.8% | 100 MG
SDP
Catalog No. 5000338189
Encompass_Preferred
PROTAC CRBN Degrader-1 (Compound 14a) is a VHL-CRBN heterodimerization PROTAC degrader. It selectively degrades CRBN by binding two E3 ubiquitin ligases (VHL and CRBN) to each other. This compound consists of a VHL ligand, a CRBN ligand, and a linker, and is intended for research use only.
  • Selectively degrades CRBN.
  • Binds two E3 ubiquitin ligases (VHL and CRBN) to each other.
  • Exhibits rapid degradation of CRBN (DC50=200 nM) in HeLa cells.
  • Shows relatively weaker degrading activity against VHL proteins.
  • Consists of a VHL ligand, a CRBN ligand, and a linker.
Catalog No. 50-003-38189 Supplier Medchemexpress LLC Supplier No. HY128845100MG
PROTAC CRBN Degrader-1 (Compound 14a) is a VHL-CRBN heterodimerization PROTAC degrader. It selectively degrades CRBN by binding two E3 ubiquitin ligases (VHL and CRBN) to each other. This compound consists of a VHL ligand, a CRBN ligand, and a linker, and is intended for research use only.
  • Selectively degrades CRBN.
  • Binds two E3 ubiquitin ligases (VHL and CRBN) to each other.
  • Exhibits rapid degradation of CRBN (DC50=200 nM) in HeLa cells.
  • Shows relatively weaker degrading activity against VHL proteins.
  • Consists of a VHL ligand, a CRBN ligand, and a linker.