Learn More
Medchemexpress LLC PROTAC CRBN Degrader-1 | 2358775-70-7 | 98.8% | 50 MG
SDP
Catalog No. 5000338187
Encompass_Preferred
PROTAC CRBN Degrader-1 (Compound 14a) is a VHL-CRBN heterodimerization PROTAC degrader designed to achieve selective degradation of CRBN by binding two E3 ubiquitin ligases (VHL and CRBN) to each other.
  • Achieves selective degradation of CRBN
  • Binds two E3 ubiquitin ligases (VHL and CRBN) to each other
  • Exhibits rapid degradation of CRBN (DC50=200 nM) in HeLa cells
  • Shows relatively weak degrading activity against VHL proteins
Catalog No. 50-003-38187 Supplier Medchemexpress LLC Supplier No. HY12884550MG
PROTAC CRBN Degrader-1 (Compound 14a) is a VHL-CRBN heterodimerization PROTAC degrader designed to achieve selective degradation of CRBN by binding two E3 ubiquitin ligases (VHL and CRBN) to each other.
  • Achieves selective degradation of CRBN
  • Binds two E3 ubiquitin ligases (VHL and CRBN) to each other
  • Exhibits rapid degradation of CRBN (DC50=200 nM) in HeLa cells
  • Shows relatively weak degrading activity against VHL proteins