Learn More
Cayman Chemical PROTAC CRBN Degrader-1 5mg
SDP
Catalog No. 502945572
Encompass_Preferred
A PROTAC that drives CRBN degradation (DC50 = 200 nM in HeLa cells)
Catalog No. 50-294-5572 Supplier Cayman Chemical Supplier No. 418785 MG
A PROTAC that drives CRBN degradation (DC50 = 200 nM in HeLa cells)