Learn More
Cell Signaling Technology PROTAC E3 Ligase Profiling Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502729502
Encompass
The PROTAC E3 Ligase Profiling Antibody Sampler Kit provides an economical solution for the detection of selected E3 ligases frequently involved in proteolysis-targeting chimera (PROTAC) research. Each kit contains a collection of primary antibodies optimized to detect key E3 ligases used in targeted protein degradation studies, along with an HRP-linked secondary antibody, facilitating efficient profiling via western blot analysis.
  • Includes seven primary antibodies targeting VHL, c-IAP1, CRBN, MDM2, KEAP1, XIAP, and β-Trcp.
  • Supplied with one Anti-rabbit IgG, HRP-linked secondary antibody.
  • Provides sufficient reagent volume to perform two western blot experiments per primary antibody.
  • Validated for multiple applications including western blotting, immunoprecipitation, and immunohistochemistry.
  • Supplied in a stabilized buffer containing 50% glycerol for storage at -20°C.
Catalog No. 50-272-9502 Supplier Cell Signaling Technology Supplier No. 99253T
The PROTAC E3 Ligase Profiling Antibody Sampler Kit provides an economical solution for the detection of selected E3 ligases frequently involved in proteolysis-targeting chimera (PROTAC) research. Each kit contains a collection of primary antibodies optimized to detect key E3 ligases used in targeted protein degradation studies, along with an HRP-linked secondary antibody, facilitating efficient profiling via western blot analysis.
  • Includes seven primary antibodies targeting VHL, c-IAP1, CRBN, MDM2, KEAP1, XIAP, and β-Trcp.
  • Supplied with one Anti-rabbit IgG, HRP-linked secondary antibody.
  • Provides sufficient reagent volume to perform two western blot experiments per primary antibody.
  • Validated for multiple applications including western blotting, immunoprecipitation, and immunohistochemistry.
  • Supplied in a stabilized buffer containing 50% glycerol for storage at -20°C.