Learn More
Medchemexpress LLC PROTAC EGFR degrader 3 | 2768472-28-0 | 97.4% | C60H77N13O5S | 1 MG
SDP
Catalog No. 5000355749
Encompass_Preferred
PROTAC EGFR degrader 3 is a potent PROTAC EGFR degrader. It exhibits excellent cellular activity against H1975 and HCC827 cells with high selectivity. Research indicates that the lysosome is involved in the degradation process of EGFR mutant degradation. This product is intended for research use only and not for sale to patients.
  • Potent PROTAC EGFR degrader
  • Excellent cellular activity against H1975 and HCC827 cells
  • High selectivity
  • Lysosome involved in EGFR mutant degradation process
  • For research use only
Catalog No. 50-003-55749 Supplier Medchemexpress LLC Supplier No. HY1446051MG
PROTAC EGFR degrader 3 is a potent PROTAC EGFR degrader. It exhibits excellent cellular activity against H1975 and HCC827 cells with high selectivity. Research indicates that the lysosome is involved in the degradation process of EGFR mutant degradation. This product is intended for research use only and not for sale to patients.
  • Potent PROTAC EGFR degrader
  • Excellent cellular activity against H1975 and HCC827 cells
  • High selectivity
  • Lysosome involved in EGFR mutant degradation process
  • For research use only