Learn More
Medchemexpress LLC PROTAC EGFR degrader 9 | 2992670-33-2 | 98.4% | 899.98 | 5 MG
SDP
Catalog No. 5000394572
Encompass_Preferred
PROTAC EGFR degrader 9 (Compound C6) is an orally active CRBN-based PROTAC EGFR degrader. It demonstrates a DC50 of 10.2 nM and a Kd of 240.2 nM against EGFRL858R/T790M/C797S. This degrader exhibits potent degradation activity against various EGFR mutants while sparing EGFRWT. The compound is for research use only and not sold to patients.
  • Orally active CRBN-based PROTAC EGFR degrader.
  • Exhibits potent degradation activity against various EGFR mutants (EGFRL858R/T790M/C797S, EGFRdel19 T790M C797S, EGFRL858R/T790M, EGFRdel19).
  • Spares EGFRWT.
  • Inhibits NSCLC tumor growth in vivo without obvious toxicity.
  • Reduces EGFR protein level and inhibits EGFR phosphorylation in tumor tissues.
  • Arrests H1975-TM cells in the G0/G1 phase.
  • Promotes cell apoptosis in H1975-TM cells.
Catalog No. 50-003-94572 Supplier Medchemexpress LLC Supplier No. HY1615365MG
PROTAC EGFR degrader 9 (Compound C6) is an orally active CRBN-based PROTAC EGFR degrader. It demonstrates a DC50 of 10.2 nM and a Kd of 240.2 nM against EGFRL858R/T790M/C797S. This degrader exhibits potent degradation activity against various EGFR mutants while sparing EGFRWT. The compound is for research use only and not sold to patients.
  • Orally active CRBN-based PROTAC EGFR degrader.
  • Exhibits potent degradation activity against various EGFR mutants (EGFRL858R/T790M/C797S, EGFRdel19 T790M C797S, EGFRL858R/T790M, EGFRdel19).
  • Spares EGFRWT.
  • Inhibits NSCLC tumor growth in vivo without obvious toxicity.
  • Reduces EGFR protein level and inhibits EGFR phosphorylation in tumor tissues.
  • Arrests H1975-TM cells in the G0/G1 phase.
  • Promotes cell apoptosis in H1975-TM cells.