Learn More
Medchemexpress LLC HY-112098 2mg Medchemexpress, PROTAC ERα Degrader-1 CAS: Purity:>98%
SDP
Catalog No. 501969022
Encompass_Preferred
Medchemexpress, HY-112098 2mg PROTAC ERα Degrader-1 CAS: PROTAC ERα Degrader-1 comprises an ubiquitin E3 ligase binding group, a linker and a protein binding group. PROTAC ERα Degrader-1 extracts from patent WO2017201449A1, compound P1. PROTAC ERα Degrader-1 is an estrogen receptor-alpha (ERα) degrader. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9022 Supplier Medchemexpress LLC Supplier No. HY1120982MG
Medchemexpress, HY-112098 2mg PROTAC ERα Degrader-1 CAS: PROTAC ERα Degrader-1 comprises an ubiquitin E3 ligase binding group, a linker and a protein binding group. PROTAC ERα Degrader-1 extracts from patent WO2017201449A1, compound P1. PROTAC ERα Degrader-1 is an estrogen receptor-alpha (ERα) degrader. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.