Learn More
Medchemexpress LLC PROTAC ERα Degrader-4 | 2521299-80-7 | 99.1% | 1074.23 | 5 MG
SDP
Catalog No. 5000359537
Encompass_Preferred
PROTAC ERα Degrader-4 (Compound ZD12) is a highly potent and selective PROTAC ERα degrader with a Ki of 5.08 μM. It contains OBHSAs, linker, and E3 ligase ligands, demonstrating excellent cell inhibitory and ERα degradation activity against Tamoxifen-sensitive and -resistant ER+ breast cancer (BC) cells and ERα-mutated BC cells. It can induce apoptosis and is suitable for cancer research.
  • Highly potent and selective ERα degrader
  • Contains OBHSAs, linker, and E3 ligase ligands
  • Exhibits excellent cell inhibitory and ERα degradation activity
  • Effective against Tamoxifen-sensitive and -resistant ER+ breast cancer (BC) cells
  • Effective against ERα-mutated BC cells
  • Induces apoptosis
  • Suitable for cancer research
Catalog No. 50-003-59537 Supplier Medchemexpress LLC Supplier No. HY1492955MG
PROTAC ERα Degrader-4 (Compound ZD12) is a highly potent and selective PROTAC ERα degrader with a Ki of 5.08 μM. It contains OBHSAs, linker, and E3 ligase ligands, demonstrating excellent cell inhibitory and ERα degradation activity against Tamoxifen-sensitive and -resistant ER+ breast cancer (BC) cells and ERα-mutated BC cells. It can induce apoptosis and is suitable for cancer research.
  • Highly potent and selective ERα degrader
  • Contains OBHSAs, linker, and E3 ligase ligands
  • Exhibits excellent cell inhibitory and ERα degradation activity
  • Effective against Tamoxifen-sensitive and -resistant ER+ breast cancer (BC) cells
  • Effective against ERα-mutated BC cells
  • Induces apoptosis
  • Suitable for cancer research