Learn More
Medchemexpress LLC Protac errα degrader-3 | 2306388-65-6 | 98.8% | 956.99 | 10 MM * 1 ML
SDP
Catalog No. 5000348387
Encompass_Preferred
PROTAC ERRα Degrader-3 is a potent and selective ERRα degrader based on von Hippel-Lindau ligand, capable of specifically degrading ERRα protein by >80% at 30 nM and is inactive against ERRβ and ERRγ proteins.
  • Dose-dependently induces ERRα degradation.
  • Decreases protein levels of ERRα downstream target genes.
  • Exhibits an IC50 value of 12.67 nM to block protein-protein interaction of ERRα with PGC-1α peptide.
Catalog No. 50-003-48387 Supplier Medchemexpress LLC Supplier No. HY13918510MM/1ML
PROTAC ERRα Degrader-3 is a potent and selective ERRα degrader based on von Hippel-Lindau ligand, capable of specifically degrading ERRα protein by >80% at 30 nM and is inactive against ERRβ and ERRγ proteins.
  • Dose-dependently induces ERRα degradation.
  • Decreases protein levels of ERRα downstream target genes.
  • Exhibits an IC50 value of 12.67 nM to block protein-protein interaction of ERRα with PGC-1α peptide.