Learn More
Medchemexpress LLC Protac ERRα degrader-3 | 2306388-65-6 | 98.8% | C47H50F6N6O7S | 10MG
SDP
Catalog No. 5000260890
Encompass_Preferred
PROTAC ERRα Degrader-3 is a proteolysis-targeting chimera (PROTAC) that selectively induces degradation of estrogen-related receptor alpha (ERRα). It is built on a von Hippel-Lindau (VHL) E3 ligase ligand and is reported to degrade ERRα by more than 80% at 30 nM while showing no activity against ERRβ or ERRγ.
  • Selective degradation of ERRα (>80% at 30 nM).
  • Inactive against ERRβ and ERRγ, supporting target specificity.
  • VHL-based E3 ligase recruitment mechanism.
  • High chemical purity (≈98.8%).
  • Molecular formula C47H50F6N6O7S; molecular weight 956.99.
Catalog No. 50-002-60890 Supplier Medchemexpress LLC Supplier No. HY13918510MG
PROTAC ERRα Degrader-3 is a proteolysis-targeting chimera (PROTAC) that selectively induces degradation of estrogen-related receptor alpha (ERRα). It is built on a von Hippel-Lindau (VHL) E3 ligase ligand and is reported to degrade ERRα by more than 80% at 30 nM while showing no activity against ERRβ or ERRγ.
  • Selective degradation of ERRα (>80% at 30 nM).
  • Inactive against ERRβ and ERRγ, supporting target specificity.
  • VHL-based E3 ligase recruitment mechanism.
  • High chemical purity (≈98.8%).
  • Molecular formula C47H50F6N6O7S; molecular weight 956.99.