Learn More
Medchemexpress LLC HY-126351 5mg Medchemexpress, PROTAC ERRα ligand 2 CAS:2306388-57-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1263515MG
Encompass_Preferred
Medchemexpress, HY-126351 5mg PROTAC ERRα ligand 2 CAS:2306388-57-6 PROTAC ERRα ligand 2 is an estrogen-related receptor α (ERRα) inverse agonist with an IC50 of 5.67 nM. PROTAC ERRα ligand 2 (IC50=5.67 nM) displays a ~11-fold improved potency than XCT790 (IC50=61.3 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9726